Assay Details
Target Gene Details
Entrez Gene ID: | 6662 |
Gene Name: | SRY-box 9 |
Gene Aliases: |
CMD1, CMPD1, SRA1, SRXX2, SRXY10 |
Location: |
Chr.17:72121020-72126420 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SOX9 | NM_000346.3 | 3 | 2507 | NP_000337.1 |
| BC056420.1 | AAH56420.1 | |||
| Z46629.1 | 3 | 2494 | CAA86598.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833530 | Chr.17:72033949 - 72232973 on Build GRCh38 | Gain+Loss |
|
| esv3573809 | Chr.17:71977779 - 72157070 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map