Assay Details
Target Gene Details
Entrez Gene ID: | 64780 |
Gene Name: | microtubule associated monooxygenase, calponin and LIM domain containing 1 |
Gene Aliases: |
MICAL, MICAL-1, NICAL |
Location: |
Chr.6:109444062-109465968 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MICAL1 | NM_001286613.1 | NP_001273542.1 | ||
| AK296284.1 |
Target Gene Details
Entrez Gene ID: | 9841 |
Gene Name: | zinc finger and BTB domain containing 24 |
Gene Aliases: |
BIF1, ICF2, PATZ2, ZNF450 |
Location: |
Chr.6:109462516-109483237 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZBTB24 | NM_014797.2 | 7 | 3667 | NP_055612.2 |
| AB007901.1 | 7 | 3667 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3610337 | Chr.6:109450093 - 109465041 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map