Assay Details
Target Gene Details
Entrez Gene ID: | 100188893 |
Gene Name: | translocase of outer mitochondrial membrane 6 |
Gene Aliases: |
OBTP, TOM6 |
Location: |
Chr.6:41787443-41789896 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TOMM6 | NM_001134493.1 | NP_001127965.1 | ||
| AF216754.1 | AAF25683.1 | |||
| AJ420506.1 | ||||
| BC015975.1 | AAH15975.1 | |||
| BC018624.1 | AAH18624.1 | |||
| BC022274.1 | AAH22274.1 | |||
| BM783950.1 | ||||
| L34839.1 |
More Information
Set Membership: |
|

Genomic Map