Assay Details
Target Gene Details
Entrez Gene ID: | 9541 |
Gene Name: | corepressor interacting with RBPJ, 1 |
Gene Aliases: |
CIR |
Location: |
Chr.2:174348150-174395715 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CIR1 | NM_004882.3 | 10 | 1311 | NP_004873.3 |
| AF098297.1 | 10 | 1218 | AAD05243.1 | |
| AK292389.1 | 10 | 1294 | ||
| BC031018.1 | 10 | 1242 | ||
| BC046098.2 | 10 | 1262 | AAH46098.1 | |
| U03644.1 | 10 | 1253 | AAA17853.1 |
More Information
Set Membership: |
|

Genomic Map