Assay Details
Target Gene Details
Entrez Gene ID: | 4016 |
Gene Name: | lysyl oxidase like 1 |
Gene Aliases: |
LOL, LOXL |
Location: |
Chr.15:73926451-73952141 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOXL1 | NM_005576.3 | NP_005567.2 | ||
| XM_017022179.1 | XP_016877668.1 | |||
| AK303982.1 | ||||
| AK314222.1 | ||||
| BC015090.2 | AAH15090.1 | |||
| BC068542.1 | AAH68542.1 | |||
| L21186.1 | AAA50162.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3581762 | Chr.15:73944743 - 73947178 on Build GRCh38 | Loss |
|
| nsv428308 | Chr.15:73922773 - 74172575 on Build GRCh38 | Gain+Loss |
|
| nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map