Assay Details
Target Gene Details
Entrez Gene ID: | 414245 |
Gene Name: | DNAJC9 antisense RNA 1 |
Gene Aliases: |
C10orf103, bA537A6.3 |
Location: |
Chr.10:73247367-73276984 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJC9-AS1 | NR_038373.1 |
Target Gene Details
Entrez Gene ID: | 51021 |
Gene Name: | mitochondrial ribosomal protein S16 |
Gene Aliases: |
CGI-132, COXPD2, MRP-S16, RPMS16 |
Location: |
Chr.10:73248843-73252693 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPS16 | NM_016065.3 | NP_057149.1 | ||
| XM_017016305.1 | XP_016871794.1 | |||
| AB049948.1 | BAB41001.1 | |||
| AF151890.1 | AAD34127.1 | |||
| AK025715.1 | ||||
| AK098291.1 | ||||
| BC021106.1 | AAH21106.1 |
More Information
Set Membership: |
|

Genomic Map