Assay Details
Target Gene Details
Entrez Gene ID: | 160492 |
Gene Name: | lamin tail domain containing 1 |
Gene Aliases: |
IFLTD1, PAS1C1 |
Location: |
Chr.12:25476082-25648604 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 20 - Exon 20 |
More Information
Set Membership: |
|

Genomic Map