Assay Details
Target Gene Details
Entrez Gene ID: | 4254 |
Gene Name: | KIT ligand |
Gene Aliases: |
DCUA, DFNA69, FPH2, FPHH, KL-1, Kitl, MGF, SCF, SF, SHEP7 |
Location: |
Chr.12:88492793-88580473 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KITLG | NM_000899.4 | 10 | 5174 | NP_000890.1 |
| NM_003994.5 | 9 | 5090 | NP_003985.2 | |
| AK025245.1 | 1 | 1581 | ||
| AK055903.1 | 1 | 2584 | ||
| CR749222.1 | 9 | 4922 | CAH18078.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv483111 | Chr.12:86306223 - 88606223 on Build GRCh38 | Loss |
|
| nsv559704 | Chr.12:88372790 - 88645023 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map