Assay Details
Target Gene Details
Entrez Gene ID: | 55035 |
Gene Name: | nucleolar protein 8 |
Gene Aliases: |
C9orf34, NOP132, bA62C3.3, bA62C3.4 |
Location: |
Chr.9:92297358-92325594 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NOL8 | NM_001256394.1 | 18 | 4245 | NP_001243323.1 |
| NM_017948.5 | 17 | 4141 | NP_060418.4 | |
| NR_046106.1 | 17 | 4935 | ||
| XM_006717166.3 | 17 | 4007 | XP_006717229.1 | |
| XM_006717167.3 | 17 | 4129 | XP_006717230.1 | |
| XM_006717168.3 | 16 | 3786 | XP_006717231.1 | |
| XM_006717169.3 | 18 | 4160 | XP_006717232.1 | |
| XM_006717170.3 | 18 | 4028 | XP_006717233.1 | |
| XM_006717172.3 | 18 | 4352 | XP_006717235.1 | |
| XM_006717173.3 | 19 | 4465 | XP_006717236.1 | |
| XM_011518824.2 | 16 | 3725 | XP_011517126.1 | |
| XM_011518827.2 | 15 | 3769 | XP_011517129.1 | |
| XM_017014876.1 | 16 | 3893 | XP_016870365.1 | |
| XM_017014877.1 | 17 | 3914 | XP_016870366.1 | |
| XM_017014878.1 | 18 | 4478 | XP_016870367.1 | |
| XM_017014879.1 | 18 | 4307 | XP_016870368.1 | |
| XM_017014880.1 | 17 | 3993 | XP_016870369.1 | |
| AB105104.1 | 17 | 4107 | BAD12268.1 | |
| AK000743.1 | BAA91356.1 | |||
| AK024245.1 | 11 | 2899 | BAB14857.1 | |
| AK024786.1 | BAB15003.1 | |||
| AK225322.1 | ||||
| BC013788.1 | AAH13788.2 | |||
| BC146810.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1045165 | Chr.9:92295180 - 92376282 on Build GRCh38 | Loss |
|
| nsv1046733 | Chr.9:91928340 - 92376282 on Build GRCh38 | Gain |
|
| esv3891693 | Chr.9:92163398 - 92538277 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map