Assay Details
Target Gene Details
Entrez Gene ID: | 7352 |
Gene Name: | uncoupling protein 3 |
Gene Aliases: |
SLC25A9 |
Location: |
Chr.11:74000281-74009237 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UCP3 | NM_003356.3 | 7 | 1320 | NP_003347.1 |
| AF001787.1 | AAC51369.1 | |||
| AF011449.1 | AAC51767.1 | |||
| BC008392.1 | 5 | 859 | AAH08392.1 | |
| BU618868.1 | 1 | 350 | ||
| KU178371.1 | ||||
| KU178372.1 | ||||
| U84763.1 | 7 | 1118 | AAC51367.1 |
More Information
Set Membership: |
|

Genomic Map