Assay Details
Target Gene Details
Entrez Gene ID: | 5931 |
Gene Name: | RB binding protein 7, chromatin remodeling factor |
Gene Aliases: |
RbAp46 |
Location: |
Chr.X:16844650-16870411 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RBBP7 | NM_001198719.1 | 12 | 1926 | NP_001185648.1 |
| NM_002893.3 | 12 | 1938 | NP_002884.1 | |
| AK091911.1 | 12 | 1923 | ||
| AK127332.1 | 3 | 3951 | ||
| BQ775345.1 | 1 | 93 | ||
| CN414343.1 | 1 | 78 | ||
| U35143.1 | 12 | 1841 | AAC50231.1 | |
| X72841.1 | 12 | 1865 | CAA51360.1 |
More Information
Set Membership: |
|

Genomic Map