Assay Details
Target Gene Details
Entrez Gene ID: | 22875 |
Gene Name: | ectonucleotide pyrophosphatase/phosphodiesterase 4 (putative) |
Gene Aliases: |
NPP4 |
Location: |
Chr.6:46129964-46146699 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ENPP4 | NM_014936.4 | 2 | 871 | NP_055751.1 |
| AB020686.1 | 1 | 660 | ||
| AK291276.1 | 2 | 716 | ||
| BC018054.1 | 2 | 842 | AAH18054.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3576147 | Chr.6:46106919 - 46185586 on Build GRCh38 | Gain |
|
| nsv1022912 | Chr.6:46125672 - 46148712 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map