Assay Details
Target Gene Details
Entrez Gene ID: | 114926 |
Gene Name: | small integral membrane protein 19 |
Gene Aliases: |
C8orf40 |
Location: |
Chr.8:42541155-42552997 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMIM19 | NM_001135674.1 | 4 | 1055 | NP_001129146.1 |
| NM_001135675.1 | 4 | 663 | NP_001129147.1 | |
| NM_001135676.1 | 4 | 348 | NP_001129148.1 | |
| NM_138436.3 | 4 | 434 | NP_612445.2 | |
| XM_005273398.4 | 4 | 969 | XP_005273455.1 | |
| AK311932.1 | ||||
| BC013035.2 | 4 | 396 | AAH13035.1 | |
| BI463786.1 | 4 | 353 | ||
| BI549696.1 | 4 | 668 | ||
| DA702519.1 | ||||
| DB519016.1 | 1 | 40 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1017899 | Chr.8:42510362 - 42586679 on Build GRCh38 | Gain |
|

Genomic Map