Assay Details
Target Gene Details
Entrez Gene ID: | 128338 |
Gene Name: | DNA damage regulated autophagy modulator 2 |
Gene Aliases: |
CORD21, PRO180, TMEM77, WWFQ154 |
Location: |
Chr.1:111117332-111140216 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DRAM2 | NM_178454.4 | 3 | 293 | NP_848549.3 |
| XM_005270469.2 | 4 | 351 | XP_005270526.1 | |
| XM_005270470.1 | 4 | 474 | XP_005270527.1 | |
| XM_006710361.1 | 4 | 423 | XP_006710424.1 | |
| XM_006710362.1 | 4 | 396 | XP_006710425.1 | |
| XM_011540707.1 | 4 | 292 | XP_011539009.1 | |
| XM_011540708.1 | 4 | 372 | XP_011539010.1 | |
| XM_017000320.1 | 3 | 347 | XP_016855809.1 | |
| XM_017000321.1 | 3 | 206 | XP_016855810.1 | |
| AK309557.1 | 3 | 530 | ||
| AL833098.1 | 3 | 661 | ||
| AY336747.1 | 3 | 171 | AAR02410.1 | |
| AY358492.1 | 3 | 156 | AAQ88856.1 | |
| BC047025.1 | 2 | 108 | AAH47025.3 | |
| BC091509.1 | 4 | 385 | AAH91509.1 | |
| DC339928.1 | 3 | 293 | ||
| EF710624.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv10639 | Chr.1:111126227 - 111244431 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map