Assay Details
Target Gene Details
Entrez Gene ID: | 84549 |
Gene Name: | MAK16 homolog |
Gene Aliases: |
MAK16L, RBM13 |
Location: |
Chr.8:33485167-33501260 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAK16 | NM_032509.3 | 5 | 356 | NP_115898.2 |
| AF251062.1 | 5 | 331 | AAK34952.1 | |
| AK027469.1 | 5 | 341 | BAB55134.1 | |
| AK225476.1 | 5 | 342 | ||
| AK314491.1 | 5 | 344 | ||
| BC015965.1 | 5 | 341 | ||
| BC028230.1 | 5 | 324 | AAH28230.1 | |
| BC039740.1 | 5 | 337 | AAH39740.1 | |
| BC050528.1 | 5 | 324 | AAH50528.1 | |
| BP281245.1 | 5 | 357 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3576539 | Chr.8:32789248 - 33495217 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map