Assay Details
Target Gene Details
Entrez Gene ID: | 5192 |
Gene Name: | peroxisomal biogenesis factor 10 |
Gene Aliases: |
NALD, PBD6A, PBD6B, RNF69 |
Location: |
Chr.1:2404802-2412622 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PEX10 | NM_002617.3 | 6 | 1345 | NP_002608.1 |
| NM_153818.1 | 6 | 1405 | NP_722540.1 | |
| XM_011541573.1 | 6 | 1453 | XP_011539875.1 | |
| AB013818.1 | 6 | 1345 | BAA87895.1 | |
| AF060502.1 | AAC18133.1 | |||
| AK124816.1 | 6 | 1323 | ||
| BC000543.2 | 6 | 1364 | AAH00543.1 | |
| BC018198.1 | 6 | 1327 | AAH18198.1 |
Target Gene Details
Entrez Gene ID: | 11079 |
Gene Name: | retention in endoplasmic reticulum sorting receptor 1 |
Gene Aliases: |
- |
Location: |
Chr.1:2391314-2405444 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RER1 | NM_007033.4 | NP_008964.3 | ||
| XM_005244713.4 | XP_005244770.2 | |||
| XM_006710306.2 | XP_006710369.1 | |||
| XM_011540542.1 | XP_011538844.1 | |||
| XM_011540543.1 | XP_011538845.1 | |||
| XM_017000131.1 | XP_016855620.1 | |||
| XM_017000132.1 | XP_016855621.1 | |||
| XM_017000133.1 | XP_016855622.1 | |||
| XM_017000134.1 | XP_016855623.1 | |||
| XM_017000135.1 | XP_016855624.1 | |||
| BM682824.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv951557 | Chr.1:2266062 - 2564661 on Build GRCh38 | Deletion |
|
| nsv545072 | Chr.1:2401592 - 2475448 on Build GRCh38 | Loss |
|
| nsv823697 | Chr.1:2389693 - 2405919 on Build GRCh38 | Gain |
|
| nsv470684 | Chr.1:2394570 - 2930923 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map