Assay Details
Target Gene Details
Entrez Gene ID: | 100287944 |
Gene Name: | uncharacterized LOC100287944 |
Gene Aliases: |
- |
Location: |
Chr.12:106496410-106774831 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100287944 | NR_040246.1 | |||
| DA250223.1 |
Target Gene Details
Entrez Gene ID: | 55188 |
Gene Name: | RIC8 guanine nucleotide exchange factor B |
Gene Aliases: |
RIC8, hSyn |
Location: |
Chr.12:106774285-106889316 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RIC8B | NM_018157.2 | NP_060627.2 | ||
| XM_005268998.3 | XP_005269055.1 | |||
| XM_011538527.2 | XP_011536829.1 | |||
| XM_017019548.1 | XP_016875037.1 | |||
| XM_017019549.1 | XP_016875038.1 | |||
| AK001482.1 | BAA91717.1 | |||
| AY221116.1 | AAO65923.1 | |||
| BC034220.1 | AAH34220.1 | |||
| BC132689.1 | ||||
| BC144056.1 | ||||
| DA325821.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9005 | Chr.12:106625455 - 106973352 on Build GRCh38 | Gain |
|
| nsv560090 | Chr.12:106541358 - 106830191 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map