Assay Details
Target Gene Details
Entrez Gene ID: | 55970 |
Gene Name: | G protein subunit gamma 12 |
Gene Aliases: |
- |
Location: |
Chr.1:67701466-67833779 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GNG12 | NM_018841.5 | 4 | 379 | NP_061329.3 |
| XM_017001809.1 | 4 | 546 | XP_016857298.1 | |
| XM_017001810.1 | 5 | 787 | XP_016857299.1 | |
| XM_017001811.1 | 5 | 716 | XP_016857300.1 | |
| AF119663.1 | 4 | 363 | AAF17220.1 | |
| AF123766.1 | 4 | 370 | AAP97245.1 | |
| AF365871.1 | 4 | 306 | AAK53385.1 | |
| AF493879.1 | AAM12593.1 | |||
| AK223000.1 | 4 | 321 | BAD96720.1 | |
| AK293101.1 | 5 | 320 | ||
| AL832431.1 | 4 | 332 | CAH10645.1 | |
| BC005940.1 | 4 | 360 | AAH05940.1 | |
| BP236225.1 | 4 | 366 | ||
| BP236699.1 | 4 | 379 |
More Information
Set Membership: |
|

Genomic Map