Assay Details
Target Gene Details
Entrez Gene ID: | 79157 |
Gene Name: | major facilitator superfamily domain containing 11 |
Gene Aliases: |
ET |
Location: |
Chr.17:76736565-76803805 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MFSD11 | NM_001242532.1 | 1 | 788 | NP_001229461.1 |
| NM_001242533.1 | NP_001229462.1 | |||
| NM_001242534.1 | NP_001229463.1 | |||
| NM_001242535.1 | NP_001229464.1 | |||
| NM_001242536.1 | NP_001229465.1 | |||
| NM_001242537.1 | NP_001229466.1 | |||
| NM_024311.3 | NP_077287.1 | |||
| XM_011525236.2 | 1 | 52 | XP_011523538.1 | |
| XM_011525237.2 | XP_011523539.2 | |||
| XM_011525238.2 | XP_011523540.2 | |||
| XM_011525239.2 | XP_011523541.2 | |||
| XM_011525240.2 | XP_011523542.2 | |||
| XM_011525241.2 | XP_011523543.2 | |||
| XM_011525242.1 | 1 | 52 | XP_011523544.1 | |
| XM_011525244.1 | 1 | 52 | XP_011523546.1 | |
| XM_011525247.2 | XP_011523549.2 | |||
| XM_017025065.1 | XP_016880554.1 | |||
| XM_017025066.1 | XP_016880555.1 | |||
| XM_017025067.1 | XP_016880556.1 | |||
| XM_017025068.1 | XP_016880557.1 | |||
| XM_017025069.1 | 1 | 282 | XP_016880558.1 | |
| XM_017025070.1 | 1 | 282 | XP_016880559.1 | |
| AF015183.1 | 1 | 902 | ||
| AF015184.1 | AAB92495.1 | |||
| AF015185.1 | AAB92496.1 | |||
| AF015186.1 | AAB92497.1 | |||
| AF015187.1 | ||||
| AF015188.1 | ||||
| AF015189.1 | 1 | 1978 | ||
| AF015190.1 | ||||
| AK000233.1 | 1 | 170 | BAA91025.1 | |
| AK308058.1 | ||||
| BC002753.2 | AAH02753.1 | |||
| BC062563.1 | AAH62563.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833550 | Chr.17:76712038 - 76886290 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map