Assay Details
Target Gene Details
Entrez Gene ID: | 56163 |
Gene Name: | ring finger protein 17 |
Gene Aliases: |
Mmip-2, SPATA23, TDRD4 |
Location: |
Chr.13:24747641-24888816 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF17 | NM_001184993.1 | NP_001171922.1 | ||
| NM_031277.2 | NP_112567.2 | |||
| XM_006719846.3 | XP_006719909.1 | |||
| XM_006719849.2 | XP_006719912.1 | |||
| XM_006719852.2 | XP_006719915.1 | |||
| XM_011535152.2 | XP_011533454.1 | |||
| XM_011535155.2 | XP_011533457.1 | |||
| XM_011535156.2 | XP_011533458.1 | |||
| XM_011535157.2 | XP_011533459.1 | |||
| XM_011535158.2 | XP_011533460.1 | |||
| XM_011535159.2 | XP_011533461.1 | |||
| XM_011535160.2 | XP_011533462.1 | |||
| XM_017020676.1 | XP_016876165.1 | |||
| AF285602.1 | AAK31981.1 | |||
| AF285603.1 | AAK31982.1 | |||
| AK302405.1 | ||||
| AK302435.1 |
More Information
Set Membership: |
|

Genomic Map