Assay Details
Target Gene Details
Entrez Gene ID: | 101927027 |
Gene Name: | uncharacterized LOC101927027 |
Gene Aliases: |
- |
Location: |
Chr.2:178413659-178440243 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927027 | NR_110204.1 | |||
| NR_110205.1 | ||||
| NR_110206.1 | ||||
| AL520497.3 | ||||
| BI915242.1 | ||||
| BX371397.2 | ||||
| BX538254.1 | ||||
| DT216517.1 |
Target Gene Details
Entrez Gene ID: | 8575 |
Gene Name: | protein activator of interferon induced protein kinase EIF2AK2 |
Gene Aliases: |
DYT16, HSD14, PACT, RAX |
Location: |
Chr.2:178431414-178451240 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRKRA | NM_001139517.1 | 7 | 1501 | NP_001132989.1 |
| NM_001139518.1 | 8 | 1222 | NP_001132990.1 | |
| NM_001316362.1 | 9 | 1540 | NP_001303291.1 | |
| NM_003690.4 | 8 | 1421 | NP_003681.1 | |
| XM_011512063.1 | 8 | 1352 | XP_011510365.1 | |
| XM_011512066.1 | 9 | 1577 | XP_011510368.1 | |
| XM_017005159.1 | 9 | 1462 | XP_016860648.1 | |
| AA084317.1 | ||||
| AF072860.1 | 8 | 1316 | AAC25672.1 | |
| AF083033.1 | AAD33099.1 | |||
| AK092918.1 | 1 | 1712 | ||
| AK093360.1 | 4 | 1447 | ||
| AK223107.1 | 8 | 1320 | BAD96827.1 | |
| AK290601.1 | 8 | 1347 | ||
| AK296019.1 | 9 | 1451 | ||
| AL136615.1 | CAB66550.1 | |||
| AL833867.1 | 7 | 1501 | CAD38725.1 | |
| BC009470.1 | 8 | 1327 | AAH09470.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv820287 | Chr.2:178431324 - 178432287 on Build GRCh38 | Loss |
|
| esv24835 | Chr.2:178431476 - 178432258 on Build GRCh38 | Loss |
|
| nsv821827 | Chr.2:178431413 - 178432002 on Build GRCh38 | Gain |
|
| dgv679n67 | Chr.2:178431353 - 178432377 on Build GRCh38 | Gain |
|
| nsv821160 | Chr.2:178431343 - 178432377 on Build GRCh38 | Duplication |
|
| nsv834467 | Chr.2:178393413 - 178566238 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map