Assay Details
Target Gene Details
Entrez Gene ID: | 22919 |
Gene Name: | microtubule associated protein RP/EB family member 1 |
Gene Aliases: |
EB1 |
Location: |
Chr.20:32819780-32850405 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAPRE1 | NM_012325.2 | NP_036457.1 | ||
| XM_011528696.2 | XP_011526998.1 | |||
| AK297273.1 | ||||
| AK310817.1 | ||||
| AK312590.1 | ||||
| BC106068.1 | AAI06069.1 | |||
| BC109281.3 | AAI09282.1 | |||
| BC128442.1 | ||||
| U24166.1 | AAC09471.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1056411 | Chr.20:32469234 - 33355029 on Build GRCh38 | Loss |
|
| nsv833959 | Chr.20:32829604 - 32982934 on Build GRCh38 | Loss |
|
| nsv179907 | Chr.20:32835381 - 32844456 on Build GRCh38 | Deletion |
|
| nsv833957 | Chr.20:32755316 - 32907992 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map