Assay Details
Target Gene Details
Entrez Gene ID: | 64843 |
Gene Name: | ISL LIM homeobox 2 |
Gene Aliases: |
- |
Location: |
Chr.15:76336724-76342476 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ISL2 | NM_145805.2 | 6 | 1423 | NP_665804.1 |
| XM_017022505.1 | 6 | 1544 | XP_016877994.1 | |
| AB593133.1 | 6 | 2945 | ||
| AK001022.1 | 6 | 1342 | ||
| BC011967.1 | 6 | 1329 | AAH11967.1 | |
| BC012136.1 | 6 | 1341 | AAH12136.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2749895 | Chr.15:75415580 - 76472296 on Build GRCh38 | Deletion |
|
| esv2749898 | Chr.15:75764003 - 77933523 on Build GRCh38 | Deletion |
|
| nsv827391 | Chr.15:76334230 - 76343986 on Build GRCh38 | Gain |
|
| nsv525769 | Chr.15:76340146 - 76369584 on Build GRCh38 | Loss |
|
| nsv1619 | Chr.15:76312275 - 76346029 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map