Assay Details
Target Gene Details
Entrez Gene ID: | 124222 |
Gene Name: | progestin and adipoQ receptor family member 4 |
Gene Aliases: |
- |
Location: |
Chr.16:2969245-2973489 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PAQR4 | NM_001284511.1 | NP_001271440.1 | ||
| NM_001284512.1 | NP_001271441.1 | |||
| NM_001284513.1 | NP_001271442.1 | |||
| NM_001324118.1 | NP_001311047.1 | |||
| NM_152341.4 | NP_689554.2 | |||
| AK054564.1 | BAB70758.1 | |||
| AK092173.1 | BAC03821.1 | |||
| AK291373.1 | ||||
| AK295348.1 | ||||
| AL832527.1 | ||||
| AY424282.1 | AAR08370.1 | |||
| BC033703.1 | AAH33703.1 | |||
| BQ935501.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv510673 | Chr.16:2919880 - 3043776 on Build GRCh38 | Deletion |
|
| nsv571246 | Chr.16:2942113 - 2973604 on Build GRCh38 | Loss |
|
| nsv827525 | Chr.16:2960856 - 2976333 on Build GRCh38 | Gain |
|
| nsv952906 | Chr.16:2868600 - 3028099 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map