Assay Details
Target Gene Details
Entrez Gene ID: | 79442 |
Gene Name: | leucine rich repeat containing 2 |
Gene Aliases: |
- |
Location: |
Chr.3:46515385-46580099 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRRC2 | NM_024512.4 | NP_078788.2 | ||
| XM_011534110.2 | XP_011532412.1 | |||
| AJ308569.2 | CAC33442.1 | |||
| AK057808.1 | BAB71585.1 | |||
| AK223440.1 | BAD97160.1 | |||
| AK314658.1 | ||||
| AK315636.1 | ||||
| BC029118.1 | AAH29118.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1013306 | Chr.3:46427523 - 46657048 on Build GRCh38 | Gain |
|

Genomic Map