Assay Details
Target Gene Details
Entrez Gene ID: | 5931 |
Gene Name: | RB binding protein 7, chromatin remodeling factor |
Gene Aliases: |
RbAp46 |
Location: |
Chr.X:16844650-16870411 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RBBP7 | NM_001198719.1 | 4 | 738 | NP_001185648.1 |
| NM_002893.3 | 4 | 750 | NP_002884.1 | |
| AK091911.1 | 4 | 735 | ||
| AK291567.1 | 4 | 494 | ||
| AK310309.1 | ||||
| BC114500.1 | 4 | 648 | ||
| BC114501.1 | 4 | 648 | ||
| CR541701.1 | 4 | 392 | CAG46502.1 | |
| U35143.1 | 4 | 653 | AAC50231.1 | |
| X72841.1 | 4 | 677 | CAA51360.1 |
More Information
Set Membership: |
|

Genomic Map