Assay Details
Target Gene Details
Entrez Gene ID: | 55329 |
Gene Name: | meiosis specific nuclear structural 1 |
Gene Aliases: |
SPATA40 |
Location: |
Chr.15:56428731-56465137 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MNS1 | NM_018365.2 | 10 | 1710 | NP_060835.1 |
| AK002084.1 | 10 | 1710 | BAA92077.1 | |
| AK057542.1 | 9 | 1510 | ||
| BC031046.1 | 10 | 1623 | AAH31046.1 | |
| BC034991.2 | 10 | 1677 | AAH34991.1 |
Target Gene Details
Entrez Gene ID: | 374618 |
Gene Name: | testis expressed 9 |
Gene Aliases: |
- |
Location: |
Chr.15:56243971-56445997 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TEX9 | NM_001286449.1 | NP_001273378.1 | ||
| NM_198524.2 | NP_940926.1 | |||
| XM_005254359.4 | XP_005254416.1 | |||
| XM_005254361.3 | 11 | 1965 | XP_005254418.1 | |
| XM_011521530.2 | XP_011519832.1 | |||
| XM_017022160.1 | 12 | 1973 | XP_016877649.1 | |
| XM_017022161.1 | XP_016877650.1 | |||
| XM_017022162.1 | 11 | 1717 | XP_016877651.1 | |
| XM_017022163.1 | 11 | 2065 | XP_016877652.1 | |
| XM_017022164.1 | 12 | 2024 | XP_016877653.1 | |
| XM_017022166.1 | XP_016877655.1 | |||
| XM_017022167.1 | 11 | 1889 | XP_016877656.1 | |
| AK294960.1 | ||||
| BC028119.1 | AAH28119.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv510662 | Chr.15:56394155 - 56443349 on Build GRCh38 | Deletion |
|
| nsv1545 | Chr.15:56367300 - 56605183 on Build GRCh38 | Deletion |
|
| esv3892699 | Chr.15:56259549 - 56692817 on Build GRCh38 | Gain |
|
| nsv498847 | Chr.15:56374840 - 56604197 on Build GRCh38 | Loss |
|
| esv35150 | Chr.15:56385532 - 56604106 on Build GRCh38 | Loss |
|
| dgv10n31 | Chr.15:56367511 - 56604187 on Build GRCh38 | Loss |
|
| nsv9270 | Chr.15:56394748 - 56468222 on Build GRCh38 | Loss |
|
| nsv433294 | Chr.15:56384259 - 56561337 on Build GRCh38 | Loss |
|
| esv2525559 | Chr.15:56377096 - 56605214 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map