Assay Details
Target Gene Details
Entrez Gene ID: | 23334 |
Gene Name: | seizure threshold 2 homolog (mouse) |
Gene Aliases: |
C1orf84, EIEE18, KIAA0467, SZT2A, SZT2B |
Location: |
Chr.1:43389884-43454247 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SZT2 | NM_015284.3 | NP_056099.3 | ||
| XM_005270686.3 | XP_005270743.1 | |||
| XM_006710501.3 | XP_006710564.1 | |||
| XM_011541106.2 | XP_011539408.1 | |||
| XM_011541107.2 | XP_011539409.1 | |||
| XM_017000819.1 | XP_016856308.1 | |||
| XM_017000820.1 | XP_016856309.1 | |||
| XM_017000821.1 | XP_016856310.1 | |||
| AK091821.1 | 6 | 1626 | BAC03755.1 | |
| BU101724.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv546128 | Chr.1:43301467 - 43451738 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map