Assay Details
Target Gene Details
Entrez Gene ID: | 1087 |
Gene Name: | carcinoembryonic antigen related cell adhesion molecule 7 |
Gene Aliases: |
CGM2 |
Location: |
Chr.19:41673307-41688277 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CEACAM7 | NM_001291485.1 | 3 | 594 | NP_001278414.1 |
| NM_006890.4 | 3 | 594 | NP_008821.2 | |
| AF006622.1 | 3 | 485 | AAB62924.1 | |
| AF006623.1 | AAB62925.1 | |||
| AK292213.1 | 3 | 587 | ||
| AK296634.1 | 4 | 524 | ||
| AK311285.1 | 3 | 422 | ||
| AK313108.1 | 3 | 587 | ||
| BC121132.2 | 3 | 488 | ||
| DC345133.1 | ||||
| L31792.1 | 3 | 484 | AAA66186.1 | |
| X98311.1 | 3 | 484 | CAA66955.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1061309 | Chr.19:41683785 - 41724525 on Build GRCh38 | Gain |
|
| esv2718577 | Chr.19:41576456 - 41709018 on Build GRCh38 | Deletion |
|
| esv29931 | Chr.19:41682561 - 41687778 on Build GRCh38 | Gain |
|
| nsv442464 | Chr.19:41683409 - 41687366 on Build GRCh38 | Gain |
|
| esv3644391 | Chr.19:41682404 - 41687675 on Build GRCh38 | Gain |
|
| nsv960841 | Chr.19:41683363 - 41684362 on Build GRCh38 | Duplication |
|
| nsv522854 | Chr.19:41613927 - 42426852 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map