Assay Details
Target Gene Details
Entrez Gene ID: | 10745 |
Gene Name: | putative homeodomain transcription factor 1 |
Gene Aliases: |
PHTF |
Location: |
Chr.1:113697202-113759888 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PHTF1 | NM_001323041.1 | 19 | 3092 | NP_001309970.1 |
| NM_001323042.1 | 19 | 2576 | NP_001309971.1 | |
| NM_001323043.1 | 19 | 2977 | NP_001309972.1 | |
| NM_001323044.1 | 19 | 3350 | NP_001309973.1 | |
| NM_001323045.1 | 20 | 3061 | NP_001309974.1 | |
| NM_001323046.1 | 16 | 2588 | NP_001309975.1 | |
| NM_006608.2 | 18 | 2881 | NP_006599.2 | |
| NR_136564.1 | 21 | 3227 | ||
| NR_136565.1 | 19 | 2868 | ||
| NR_136566.1 | 20 | 3143 | ||
| NR_136567.1 | 20 | 3028 | ||
| AJ011863.1 | 18 | 2460 | CAB51572.1 | |
| AK126773.1 | 12 | 3580 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3587240 | Chr.1:113533246 - 114158910 on Build GRCh38 | Loss |
|
| esv3587239 | Chr.1:113510852 - 113866662 on Build GRCh38 | Loss |
|
| esv23869 | Chr.1:113320330 - 114358495 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map