Assay Details
Target Gene Details
Entrez Gene ID: | 55035 |
Gene Name: | nucleolar protein 8 |
Gene Aliases: |
C9orf34, NOP132, bA62C3.3, bA62C3.4 |
Location: |
Chr.9:92297358-92325594 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 16 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1045165 | Chr.9:92295180 - 92376282 on Build GRCh38 | Loss |
|
| nsv1046733 | Chr.9:91928340 - 92376282 on Build GRCh38 | Gain |
|
| esv3891693 | Chr.9:92163398 - 92538277 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map