Assay Details
Target Gene Details
Entrez Gene ID: | 10125 |
Gene Name: | RAS guanyl releasing protein 1 |
Gene Aliases: |
CALDAG-GEFI, CALDAG-GEFII, RASGRP, V, hRasGRP1 |
Location: |
Chr.15:38488097-38564806 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RASGRP1 | NM_001128602.1 | 16 | 3959 | NP_001122074.1 |
| NM_001306086.1 | 15 | 3573 | NP_001293015.1 | |
| NM_005739.3 | 17 | 4064 | NP_005730.2 | |
| XM_005254114.2 | 16 | 3900 | XP_005254171.1 | |
| XM_011521151.2 | 16 | 3667 | XP_011519453.1 | |
| XM_017021860.1 | 17 | 3991 | XP_016877349.1 | |
| AB208848.1 | 10 | 3085 | BAD92085.1 | |
| AF081195.1 | 17 | 3989 | AAC79699.1 | |
| AF081197.1 | 16 | 3843 | AAF21898.1 |
More Information
Set Membership: |
|

Genomic Map