Assay Details
Target Gene Details
Entrez Gene ID: | 53938 |
Gene Name: | peptidylprolyl isomerase like 3 |
Gene Aliases: |
CYPJ |
Location: |
Chr.2:200870907-200889333 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
More Information
Set Membership: |
|

Genomic Map