Assay Details
Target Gene Details
Entrez Gene ID: | 8836 |
Gene Name: | gamma-glutamyl hydrolase |
Gene Aliases: |
GH |
Location: |
Chr.8:63015079-63039051 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GGH | NM_003878.2 | 6 | 828 | NP_003869.1 |
| XM_011517623.2 | 6 | 579 | XP_011515925.1 | |
| AK290450.1 | 6 | 572 | ||
| AK301094.1 | 6 | 578 | ||
| BC025025.1 | 6 | 629 | AAH25025.1 | |
| U55206.1 | 6 | 604 | AAC05579.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1018879 | Chr.8:62369165 - 63025073 on Build GRCh38 | Gain |
|
| nsv6227 | Chr.8:62979528 - 63024393 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map