Assay Details
Target Gene Details
Entrez Gene ID: | 29080 |
Gene Name: | coiled-coil domain containing 59 |
Gene Aliases: |
BR22, HSPC128, TAP26 |
Location: |
Chr.12:82352304-82358805 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC59 | NM_014167.4 | NP_054886.2 | ||
| NR_033192.1 | ||||
| AF161477.1 | AAF29092.1 | |||
| AF213377.1 | AAG43569.1 | |||
| AL832749.1 | ||||
| BC020647.1 | AAH20647.1 | |||
| BG538660.1 | ||||
| BM846580.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv559539 | Chr.12:81887018 - 82502439 on Build GRCh38 | Gain |
|
| nsv832471 | Chr.12:82185174 - 82361581 on Build GRCh38 | Loss |
|
| nsv559541 | Chr.12:82321431 - 82714616 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map