Assay Details
Target Gene Details
Entrez Gene ID: | 5903 |
Gene Name: | RAN binding protein 2 |
Gene Aliases: |
ADANE, ANE1, IIAE3, NUP358, TRP1, TRP2 |
Location: |
Chr.2:108719446-109842301 on Build GRCh38 |
Assay Gene Location: | Within Intron 32 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RANBP2 | XM_017004623.1 | XP_016860112.1 |
Target Gene Details
Entrez Gene ID: | 151011 |
Gene Name: | septin 10 |
Gene Aliases: |
- |
Location: |
Chr.2:109542797-109614247 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 11 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2874 | Chr.2:109531796 - 109576044 on Build GRCh38 | Deletion |
|
| dgv1162e212 | Chr.2:109504090 - 110630891 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map