Assay Details
Target Gene Details
Entrez Gene ID: | 6705 |
Gene Name: | small proline rich protein 2F |
Gene Aliases: |
- |
Location: |
Chr.1:153112121-153119460 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPRR2F | XM_017002176.1 | XP_016857665.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv946396 | Chr.1:153099343 - 153115862 on Build GRCh38 | Duplication |
|
| nsv547920 | Chr.1:153060930 - 153126869 on Build GRCh38 | Loss |
|
| esv3587575 | Chr.1:153096350 - 153114772 on Build GRCh38 | Gain |
|
| esv22397 | Chr.1:153096719 - 153115487 on Build GRCh38 | Gain |
|
| nsv547899 | Chr.1:152699529 - 153272606 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map