Assay Details
Target Gene Details
Entrez Gene ID: | 7159 |
Gene Name: | tumor protein p53 binding protein 2 |
Gene Aliases: |
53BP2, ASPP2, BBP, P53BP2, PPP1R13A |
Location: |
Chr.1:223779893-223845981 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TP53BP2 | NM_001031685.2 | NP_001026855.2 | ||
| NM_005426.2 | NP_005417.1 | |||
| AA961496.1 | ||||
| AI042183.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4698 | Chr.1:223770507 - 223787519 on Build GRCh38 | Insertion |
|
| esv34082 | Chr.1:223663316 - 223956807 on Build GRCh38 | Loss |
|
| esv2759000 | Chr.1:223747518 - 224175461 on Build GRCh38 | Gain+Loss |
|
| nsv832681 | Chr.1:223524706 - 223814989 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map