Assay Details
Target Gene Details
Entrez Gene ID: | 64101 |
Gene Name: | leucine rich repeat containing 4 |
Gene Aliases: |
NAG14, NGL-2 |
Location: |
Chr.7:128027071-128032273 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRRC4 | NM_022143.4 | 2 | 2713 | NP_071426.1 |
| XM_011516461.2 | 3 | 3019 | XP_011514763.1 | |
| XM_017012505.1 | 3 | 2909 | XP_016867994.1 | |
| AF196976.4 | 2 | 2716 | AAG28019.2 | |
| BC111561.1 | 2 | 2707 | ||
| BC111745.1 | 2 | 2707 |
Target Gene Details
Entrez Gene ID: | 27044 |
Gene Name: | staphylococcal nuclease and tudor domain containing 1 |
Gene Aliases: |
TDRD11, p100 |
Location: |
Chr.7:127652119-128092607 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SND1 | NM_014390.2 | NP_055205.2 | ||
| AB209510.1 | BAD92747.1 | |||
| AK096583.1 | ||||
| AK304181.1 | ||||
| AK309014.1 | ||||
| AK312313.1 | ||||
| BC017180.2 | ||||
| BE799345.1 | ||||
| BT009785.1 | AAP88787.1 | |||
| U22055.1 | AAA80488.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv5944 | Chr.7:128024907 - 128058792 on Build GRCh38 | Insertion |
|
| nsv1024743 | Chr.7:127858642 - 128029951 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map