Assay Details
Target Gene Details
Entrez Gene ID: | 64854 |
Gene Name: | ubiquitin specific peptidase 46 |
Gene Aliases: |
- |
Location: |
Chr.4:52590960-52659335 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP46 | NM_001134223.1 | 9 | 3103 | NP_001127695.1 |
| NM_001286767.1 | 9 | 2991 | NP_001273696.1 | |
| NM_001286768.1 | 10 | 3194 | NP_001273697.1 | |
| NM_022832.3 | 9 | 3027 | NP_073743.2 | |
| XM_017008557.1 | 9 | 3052 | XP_016864046.1 | |
| AK024318.1 | 9 | 2968 | BAB14881.1 | |
| BC037574.1 | 9 | 2976 | AAH37574.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524494 | Chr.4:52593454 - 52715898 on Build GRCh38 | Loss |
|
| nsv594201 | Chr.4:51932458 - 52866278 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map