Assay Details
Target Gene Details
Entrez Gene ID: | 23398 |
Gene Name: | peptidylprolyl isomerase domain and WD repeat containing 1 |
Gene Aliases: |
- |
Location: |
Chr.5:65563236-65587549 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPWD1 | NM_001278926.1 | 12 | 1983 | NP_001265855.1 |
| NM_001278927.1 | 11 | 1773 | NP_001265856.1 | |
| NM_001278929.1 | 10 | 1843 | NP_001265858.1 | |
| NM_015342.3 | 11 | 1949 | NP_056157.1 | |
| XM_005248474.3 | 7 | 1116 | XP_005248531.1 | |
| XM_006714583.3 | 12 | 1919 | XP_006714646.1 | |
| XM_011543295.1 | 10 | 2231 | XP_011541597.1 | |
| XM_017009286.1 | 10 | 1273 | XP_016864775.1 | |
| AK025679.1 | 11 | 1896 | ||
| AK057718.1 | 1 | 2505 | ||
| AK298175.1 | 11 | 1713 | ||
| AK300022.1 | 12 | 1926 | ||
| AK301654.1 | 10 | 1783 | ||
| BC015385.1 | 11 | 1877 | AAH15385.1 | |
| BM669378.1 | 1 | 238 | ||
| D38552.1 | 11 | 1872 | BAA07555.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3894204 | Chr.5:65586109 - 65714937 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map