Assay Details
Target Gene Details
Entrez Gene ID: | 30 |
Gene Name: | acetyl-CoA acyltransferase 1 |
Gene Aliases: |
ACAA, PTHIO, THIO |
Location: |
Chr.3:38122710-38137242 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACAA1 | NM_001130410.1 | NP_001123882.1 | ||
| NM_001607.3 | NP_001598.1 | |||
| NR_024024.1 | ||||
| XM_006713122.1 | XP_006713185.1 | |||
| XM_006713123.2 | XP_006713186.1 | |||
| XM_011533650.2 | XP_011531952.1 | |||
| AF035295.1 | AAB88181.1 | |||
| AK025138.1 | ||||
| AK127051.1 | ||||
| BC000635.2 | AAH00635.1 | |||
| BC011977.1 | AAH11977.1 | |||
| BC014474.1 | AAH14474.1 | |||
| X14813.1 | CAA32918.1 | |||
| X51460.1 | CAA35825.1 |
Target Gene Details
Entrez Gene ID: | 9940 |
Gene Name: | deleted in lung and esophageal cancer 1 |
Gene Aliases: |
CFAP81, DLC-1, DLC1, F56 |
Location: |
Chr.3:38038595-38122741 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 38 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DLEC1 | NM_001321153.1 | NP_001308082.1 | ||
| NM_007335.3 | NP_031361.2 | |||
| NM_007337.3 | NP_031363.2 | |||
| XM_006713438.3 | XP_006713501.1 | |||
| XM_011534317.2 | XP_011532619.1 | |||
| XM_011534318.2 | XP_011532620.1 | |||
| AB020522.1 | BAA77247.1 | |||
| AB425198.1 | ||||
| AL137282.1 | 5 | 1523 | CAB70676.1 | |
| AL137706.1 | CAB70884.2 | |||
| BU680752.1 | ||||
| CB243860.1 |
More Information
Set Membership: |
|

Genomic Map