Assay Details
Target Gene Details
Entrez Gene ID: | 493860 |
Gene Name: | coiled-coil domain containing 73 |
Gene Aliases: |
NY-SAR-79 |
Location: |
Chr.11:32602080-32794658 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC73 | NM_001008391.3 | 16 | 1766 | NP_001008392.2 |
| XM_011520139.2 | 15 | 1627 | XP_011518441.1 | |
| AK128159.1 | 7 | 985 | BAC87304.1 | |
| AK131470.1 | 16 | 1750 | BAD18615.1 |

Genomic Map