Assay Details
Target Gene Details
Entrez Gene ID: | 2078 |
Gene Name: | ERG, ETS transcription factor |
Gene Aliases: |
erg-3, p55 |
Location: |
Chr.21:38367261-38662045 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ERG | NM_001136154.1 | 12 | 3180 | NP_001129626.1 |
| NM_001136155.1 | 9 | 2793 | NP_001129627.1 | |
| NM_001243428.1 | 12 | 3203 | NP_001230357.1 | |
| NM_001243429.1 | 9 | 2767 | NP_001230358.1 | |
| NM_001243432.2 | NP_001230361.1 | |||
| NM_004449.4 | 11 | 3108 | NP_004440.1 | |
| NM_182918.3 | 10 | 3011 | NP_891548.1 | |
| XM_011529486.2 | XP_011527788.1 | |||
| XM_017028287.1 | XP_016883776.1 | |||
| XM_017028288.1 | XP_016883777.1 | |||
| XM_017028289.1 | XP_016883778.1 | |||
| AY204741.1 | AAP41719.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834098 | Chr.21:38319976 - 38473331 on Build GRCh38 | Gain |
|
| nsv3515 | Chr.21:38378923 - 38408351 on Build GRCh38 | Insertion |
|
| nsv1160712 | Chr.21:38379214 - 38396352 on Build GRCh38 | Deletion |
|
| nsv1058886 | Chr.21:37969257 - 38427716 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map