Assay Details
Target Gene Details
Entrez Gene ID: | 571 |
Gene Name: | BTB domain and CNC homolog 1 |
Gene Aliases: |
BACH-1, BTBD24 |
Location: |
Chr.21:29298795-29361896 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BACH1 | NM_001186.3 | 2 | 344 | NP_001177.1 |
| NM_206866.2 | 2 | 368 | NP_996749.1 | |
| NR_027655.2 | 2 | 304 | ||
| AA402241.1 | 2 | 265 | ||
| AB002803.1 | 2 | 243 | BAA24932.1 | |
| AF026199.1 | 2 | 368 | AAB84100.1 | |
| AK291021.1 | 2 | 301 | ||
| AK313791.1 | 2 | 217 | ||
| BC063307.1 | 2 | 217 | AAH63307.1 | |
| CR450303.1 | 1 | 125 | CAG29299.1 | |
| DA855751.1 | 3 | 378 | ||
| HY028322.1 | 2 | 348 |
More Information
Set Membership: |
|

Genomic Map