Assay Details
Target Gene Details
Entrez Gene ID: | 29942 |
Gene Name: | purine rich element binding protein G |
Gene Aliases: |
PURG-A, PURG-B |
Location: |
Chr.8:30995802-31033715 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PURG | NM_001015508.2 | 2 | 2135 | NP_001015508.1 |
| NM_001323312.1 | 3 | 1325 | NP_001310241.1 | |
| AY077841.1 | 2 | 2135 | AAL77218.1 |
More Information
Set Membership: |
|

Genomic Map