Assay Details
Target Gene Details
Entrez Gene ID: | 150280 |
Gene Name: | HORMA domain containing 2 |
Gene Aliases: |
CT46.2 |
Location: |
Chr.22:30080069-30207425 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HORMAD2 | XM_011529914.2 | 3 | 559 | XP_011528216.1 |
| XM_011529917.2 | 3 | 418 | XP_011528219.1 | |
| XM_011529918.1 | 3 | 559 | XP_011528220.1 | |
| XM_017028621.1 | 3 | 440 | XP_016884110.1 | |
| XM_017028622.1 | 3 | 359 | XP_016884111.1 | |
| XM_017028624.1 | 3 | 559 | XP_016884113.1 | |
| XM_017028625.1 | XP_016884114.1 | |||
| AK098703.1 | 3 | 384 | BAC05388.1 | |
| BC030013.1 | 3 | 169 | AAH30013.1 | |
| BI459310.1 | 3 | 164 | ||
| CR456341.1 | 2 | 146 | CAG30227.1 | |
| HY002382.1 | 4 | 209 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv588884 | Chr.22:30098382 - 30194037 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map