Assay Details
Target Gene Details
Entrez Gene ID: | 8310 |
Gene Name: | acyl-CoA oxidase 3, pristanoyl |
Gene Aliases: |
- |
Location: |
Chr.4:8366263-8440725 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACOX3 | NM_001101667.1 | 17 | 2285 | NP_001095137.1 |
| NM_003501.2 | 18 | 2353 | NP_003492.2 | |
| XM_005248011.4 | 18 | 2406 | XP_005248068.1 | |
| XM_005248013.3 | 17 | 2338 | XP_005248070.1 | |
| XM_011513565.2 | 18 | 2365 | XP_011511867.1 | |
| XM_011513566.1 | 16 | 2163 | XP_011511868.1 | |
| AK225080.1 | 17 | 2283 | ||
| BC017053.1 | 17 | 2271 | AAH17053.1 | |
| CA306669.1 | 1 | 538 | ||
| Y11411.1 | 18 | 2331 | CAA72214.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4223 | Chr.4:8346346 - 8373558 on Build GRCh38 | Insertion |
|
| nsv470013 | Chr.4:8314211 - 8375331 on Build GRCh38 | Gain |
|
| nsv593637 | Chr.4:8352102 - 8388919 on Build GRCh38 | Loss |
|
| nsv526508 | Chr.4:8356196 - 8369827 on Build GRCh38 | Loss |
|
| nsv1002794 | Chr.4:8108356 - 8489207 on Build GRCh38 | Gain |
|
| nsv1009011 | Chr.4:8223942 - 8423104 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs115372866] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map