Assay Details
Target Gene Details
Entrez Gene ID: | 26354 |
Gene Name: | G protein nucleolar 3 |
Gene Aliases: |
C77032, E2IG3, NNP47, NS |
Location: |
Chr.3:52685920-52694497 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GNL3 | NM_014366.4 | 6 | 637 | NP_055181.3 |
| NM_206825.1 | 6 | 734 | NP_996561.1 | |
| NM_206826.1 | 6 | 587 | NP_996562.1 | |
| AF191018.1 | 6 | 634 | AAF09482.1 | |
| AK027514.1 | 6 | 638 | BAB55168.1 | |
| AK027516.1 | 6 | 487 | BAB55169.1 | |
| AK315484.1 | 6 | 550 | ||
| AY825265.1 | 5 | 464 | AAV74413.1 | |
| BC001024.2 | 6 | 485 | AAH01024.1 | |
| BF732788.1 | 6 | 587 | ||
| CF272475.1 | 6 | 506 |
More Information
Set Membership: |
|

Genomic Map