Assay Details
Target Gene Details
Entrez Gene ID: | 148581 |
Gene Name: | ubiquitin conjugating enzyme E2 U (putative) |
Gene Aliases: |
- |
Location: |
Chr.1:64202802-64267381 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBE2U | NM_152489.1 | 2 | 354 | NP_689702.1 |
| XM_005270498.4 | 2 | 1148 | XP_005270555.1 | |
| XM_006710379.3 | 2 | 289 | XP_006710442.1 | |
| XM_011540764.2 | 3 | 1089 | XP_011539066.1 | |
| BC029895.1 | 2 | 354 | AAH29895.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv546436 | Chr.1:64205389 - 64269305 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map